Anti-West Nile Virus, NS2b/NS3 Protease Mouse Monoclonal Antibody [clone: 8H153]
Lieferant: US Biological
W1018-53
USBIW1018-53EA 893 EUR
USBIW1018-53
Anti-West Nile Virus, NS2b/NS3 Protease Mouse Monoclonal Antibody [clone: 8H153]
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml
Type: Primary
Antigen: West Nile Virus, NS2b/NS3 Protease
Clonality: monoclonal
Clone: 8H153
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen