Anti-West Nile Virus, NS3 Protease Mouse Monoclonal Antibody [clone: 8H154]

Lieferant: US Biological
W1018-54
USBIW1018-54EA 802 EUR
USBIW1018-54
Anti-West Nile Virus, NS3 Protease Mouse Monoclonal Antibody [clone: 8H154]
Antikörper
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 1-2?g/ml

Type: Primary
Antigen: West Nile Virus, NS3 Protease
Clonality: monoclonal
Clone: 8H154
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR