Anti-WFS1 Rabbit Polyclonal Antibody

Lieferant: US Biological
W1023-70
USBIW1023-70EA 512 EUR
USBIW1023-70
Anti-WFS1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:2000; Immunoprecipitation: 2?l; Immunofluorescence: 1:200; Immunochemistry: 1:1000;

Type: Primary
Antigen: WFS1 (Wolfram syndrome 1 (wolframin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR