Anti-Wnt Inhibiting Factor 1 Mouse Monoclonal Antibody [clone: 8H102]
Lieferant: US Biological
W3005-75
USBIW3005-75EA 573 EUR
USBIW3005-75
Anti-Wnt Inhibiting Factor 1 Mouse Monoclonal Antibody [clone: 8H102]
Antikörper
Type: Primary
Antigen: Wnt Inhibiting Factor 1
Clonality: monoclonal
Clone: 8H102
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human