Anti-Yellow Fever Virus Mouse Monoclonal Antibody [clone: 10B2541]

Lieferant: US Biological
Y2041-02B
USBIY2041-02BEA 796 EUR
USBIY2041-02B
Anti-Yellow Fever Virus Mouse Monoclonal Antibody [clone: 10B2541]
Antikörper
Type: Primary
Antigen: Yellow Fever Virus
Clonality: monoclonal
Clone: 10B2541
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR