Anti-ZAP70 Goat Polyclonal Antibody

Lieferant: US Biological
Z0300-01F
USBIZ0300-01FEA 870 EUR
USBIZ0300-01F
Anti-ZAP70 Goat Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1.0 ?g/ml

Type: Primary
Antigen: ZAP70 (zeta-chain (TCR) associated protein kinase 70kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR