Lieferant: UVEX
8763220 8763310 8763110 8763210 8763200 8763100
UVEX8763220EA 44.3 EUR
UVEX8763220 UVEX8763310 UVEX8763110 UVEX8763210 UVEX8763200 UVEX8763100
Atemschutzmasken Einweg-Atemschutzmasken Atemschutzmasken gegen Partikel
Respirators in protection class FFP1, FFP2 and FFP3.
  • Soft material edges for greater comfort and wearer acceptance
  • Continuous textile headband ensures that the mask fits comfortably and securely
  • Adjustable nose clip ensures an excellent individual fit and secure positioning
  • Optional: exhalation valve to reduce heat and humidity in the mask
More sustainable mask family: the valve and headband attachment on all models are partly made of recycled material.
The soft material edges prevent uncomfortable pressure points. The seamless headband can be adjusted for an optimum fit. The exhalation valve with a 360° opening ensures very low breathing resistance and a cooling effect climate inside the mask. The upper face seal offers both a more secure positioning of the mask and increased wearer comfort.
Sealing lip (in the nose area for FFP1 and FFP2, all-round for FFP3) and nose clip provide maximum protection and comfort.
FFP3 preformed masks have a four-point headband attachment.
2220: additional activated carbon filter against odours, gases and vapours below threshold.