uvex silv-Air e Particulate Respirator, FPP2/FPP3
Lieferant: UVEX
8787232 8787333 8787317 8787312 8787318 8787330 8787233
UVEX8787232EA 28.4 EUR
UVEX8787232 UVEX8787333 UVEX8787317 UVEX8787312 UVEX8787318 UVEX8787330 UVEX8787233
uvex silv-Air e Particulate Respirator, FPP2/FPP3
Atemschutzmasken Einweg-Atemschutzmasken Atemschutzmasken gegen Partikel
All masks of the uvex silv-Air e family combine our unique technologies of optimum wearer comfort with extremely low breathing resistance. The products are therefore suitable for heavy work and for being worn for long periods of time. The increased size of the filter surface even enables the high-performance models to be worn for several days.
  • High-performance climazone system with three compartments for a comfortable, cool climate inside the mask
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Flexible, adjustable integrated nose clip
  • Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
  • High-quality silv-Air outer material for optimal breathability
  • Very soft, all-around sealing lip for optimal wearer comfort and secure fit
  • Nasal recess on the sealing lip ensures ideal fit for almost any nose shape
  • Particle-filtering preformed mask with exhalation valve and inhalation valves
Two extra filters with inhalation valves for lower breathing resistance and increased comfort due to easier air exchange. Combination of 3D nose area and sealing lip ensures ideal fit for almost any nose shape. D label (dolomite dust test passed): maximum breathability even with really high dust levels. R label (reusable): tested for re-usability and performance, including in the second shift.
Note: Respirators are not suitable for people with beards, sideburns or deep scars near the seals.
Standards: EN 149:2001+A1:2009.