Tubing kit for ketone-based organic solvents
Lieferant: VARIAN
9910125000 9910125100 9910125300
VARI9910125000EA 0 EUR
VARI9910125000 VARI9910125100 VARI9910125300
Tubing kit for ketone-based organic solvents
Schläuche Chromatographie-Schläuche
Organic samples, ketone based organic solvents (includes Marprene™ pump tubing (one each black/black and white/white), FEP transfer tube (spray chamber to torch), drain tubing, tubing for waste line, one pack of EzyFit connectors, nebulizer capillary tubing, connectors and connecting tubing).