Chromatography tubing, stainless steel,
Lieferant: VARIAN
CP4006 CP4061 CP4025 CP4009 CP4024 CP4026 CP4062 CP4065 CP4005 CP4007 CP4013 CP4014
VARICP4006EA 55 EUR
VARICP4006 VARICP4061 VARICP4025 VARICP4009 VARICP4024 VARICP4026 VARICP4062 VARICP4065 VARICP4005 VARICP4007 VARICP4013 VARICP4014
Chromatography tubing, stainless steel,
Schläuche Chromatographie-Schläuche
Tubing made of 316 stainless steel.
This 316 stainless steel tubing is ideal for GC applications and corrosion resistant for HPLC applications. Its maximum operating temperature is 500 °C and maximum pressure is 10000 psi (689,5 bar).