Anti-kappa chain Antibody (AMCA)

Lieferant: Vector Laboratories
CI-3060-.5
VECTCI-3060-.5EA 448 EUR
VECTCI-3060-.5
Anti-kappa chain Antibody (AMCA)
Antikörper
This is affinity-purified, ultrapure, high affinity antibody which is optimally labeled with AMCA to provide the brightest label for fluorescence microscopy.

  • Thoroughly adsorbed against serum and immunoglobulins from potentially interfering species
  • Supplied in solution
  • Excitation: 350 nm
  • Emission: 450 nm
  • Colour of fluorescence: Blue

This kappa chain specific antibody has virtually no cross-reactivity with other immunoglobulin classes or other heavy or light chains. This can be used for flow cytometry or used for tissue staining.

Type: Secondary
Antigen: Kappa
Clonality: Polyclonal
Clone:
Conjugation: AMCA
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR