Peroxidase substrate, NovaRED™
Lieferant: Vector Laboratories
SK-4800
VECTSK-4800EA 591 EUR
VECTSK-4800
Peroxidase substrate, NovaRED™
Elektrophorese-Reagenzien Western Blotting Reagents
NovaRED™ substrate produces a red reaction product and is several times more sensitive than AEC. Moreover, sections stained with the NovaRED™ substrate may be dehydrated in ethanol and cleared in organic solvents such as xylene and permanently mounted in a non-aqueous mounting medium such as VectaMount™.
  • Excellent contrast with Vector® peroxidase and alkaline phosphatase substrates
  • Compatibility with Vector® hematoxylin and Vector® methyl green counterstains
Can be used as a label alternative to DAB or as an additional colour in multiple label tissue staining.