Peroxidase substrate, NovaRED™
Lieferant: Vector Laboratories
SK-4800
VECTSK-4800EA
591
EUR
VECTSK-4800
Peroxidase substrate, NovaRED™
Elektrophorese-Reagenzien
Western Blotting Reagents
NovaRED™ substrate produces a red reaction product and is several times more sensitive than AEC. Moreover, sections stained with the NovaRED™ substrate may be dehydrated in ethanol and cleared in organic solvents such as xylene and permanently mounted in a non-aqueous mounting medium such as VectaMount™.
- Excellent contrast with Vector® peroxidase and alkaline phosphatase substrates
- Compatibility with Vector® hematoxylin and Vector® methyl green counterstains
Can be used as a label alternative to DAB or as an additional colour in multiple label tissue staining.