Anti-PAX8 Rabbit Monoclonal Antibody [clone: ZR1]

Lieferant: IMMUNOLOGIC
VWRKILM0002-C01EA 95.7 EUR
VWRKILM0002-C01 VWRKILM0002-C05 VWRKILM0002-C1
Anti-PAX8 Rabbit Monoclonal Antibody [clone: ZR1]
Antikörper
Type: Primary
Antigen: PAX8
Clonality: Monoclonal
Clone: ZR1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR