Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1]

Lieferant: IMMUNOLOGIC
ILM07211-R10 ILM07211-R25
VWRKILM07211-R10EA 0 EUR
VWRKILM07211-R10 VWRKILM07211-R25
Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1]
Antikörper
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: UCHL-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Chimpanzee, Common Marmoset
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR