Anti-ESR1 Mouse Monoclonal Antibody [clone: TE111.5D11]

Lieferant: Abcam
AB16460-50
ABCAAB16460-50EA 901 EUR
ABCAAB16460-50
Anti-ESR1 Mouse Monoclonal Antibody [clone: TE111.5D11]
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: ESR1 (estrogen receptor 1)
Clonality: monoclonal
Clone: TE111.5D11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR