Anti-IgG Goat Polyclonal Antibody (Biotin)

Lieferant: Biotium
20184 20184-1
20184.EA 359 EUR
20184. 20184-1
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR