Anti-Fluorescein Mouse Monoclonal Antibody (FITC (Fluorescein))

Lieferant: Biotium
20211 20211-1
20211.EA 385 EUR
20211. 20211-1
Anti-Fluorescein Mouse Monoclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Tag
Antigen: Fluorescein (fluorescein)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR