Anti-EAAT1/GLAST-1/SLC1A3 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-1869 NB100-1869SS
NOVUNB100-1869EA 596 EUR
NOVUNB100-1869 NOVUNB100-1869SS
Anti-EAAT1/GLAST-1/SLC1A3 Rabbit Polyclonal Antibody
Antikörper
The EAAT1 / GLAST-1 / SLC1A3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to EAAT1 / GLAST-1 / SLC1A3. This antibody reacts with human, mouse, rat. The EAAT1 / GLAST-1 / SLC1A3 Antibody has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:500

Type: Primary
Antigen: EAAT1/GLAST-1/SLC1A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR