Anti-PINK1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB100-493 NB100-493SS
NOVUNB100-493EA 583 EUR
NOVUNB100-493 NOVUNB100-493SS
Anti-PINK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PINK1 (PTEN induced putative kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR