Anti-MRP1 Mouse Monoclonal Antibody [clone: IU5C1]
Lieferant: Novus Biologicals
NB110-57131 NB110-57131SS
NOVUNB110-57131EA 554 EUR
NOVUNB110-57131 NOVUNB110-57131SS
Anti-MRP1 Mouse Monoclonal Antibody [clone: IU5C1]
Antikörper
Type: Primary
Antigen: MRP1
Clonality: monoclonal
Clone: IU5C1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse