Anti-PDZK1IP1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB400-151 NB400-151SS
NOVUNB400-151EA 572 EUR
NOVUNB400-151 NOVUNB400-151SS
Anti-PDZK1IP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PDZK1IP1 (PDZK1 interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR