Anti-Del-1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-28632 NBP1-28632SS
NOVUNBP1-28632EA 591 EUR
NOVUNBP1-28632 NOVUNBP1-28632SS
Anti-Del-1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Del-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR