Anti-TYRO3 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-28635 NBP1-28635SS
NOVUNBP1-28635EA 591 EUR
NOVUNBP1-28635 NOVUNBP1-28635SS
Anti-TYRO3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TYRO3 (TYRO3 protein tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR