Anti-SNX27 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-45283 NBP1-45283SS
NOVUNBP1-45283EA 591 EUR
NOVUNBP1-45283 NOVUNBP1-45283SS
Anti-SNX27 Rabbit Polyclonal Antibody
Antikörper
The SNX27 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNX27. This antibody reacts with human, mouse. The SNX27 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: SNX27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR