Anti-AURKA Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-51843 NBP1-51843SS
NOVUNBP1-51843EA 606 EUR
NOVUNBP1-51843 NOVUNBP1-51843SS
Anti-AURKA Rabbit Polyclonal Antibody
Antikörper
The Aurora A Antibody from Novus Biologicals is a rabbit polyclonal antibody to Aurora A. This antibody reacts with human, mouse. The Aurora A Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Simple Western

Type: Primary
Antigen: Aurora A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR