Anti-HDAC6 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-78981SS NBP1-78981
NOVUNBP1-78981SSEA 235 EUR
NOVUNBP1-78981SS NOVUNBP1-78981
Anti-HDAC6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HDAC6 (histone deacetylase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR