Anti-schistosomal Glutathione-S-transferase Goat Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: Columbia Biosciences
D3-1310-1MG D3-1310-100
COBSD3-1310-1MGEA 860 EUR
COBSD3-1310-1MG COBSD3-1310-100
Anti-schistosomal Glutathione-S-transferase Goat Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Type: Secondary
Antigen: schistosomal Glutathione-S-Transferase
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR