Anti-Genistein Mouse Monoclonal Antibody [clone: 5G12]

Lieferant: US Biological
G2024-76-100 G2024-76-50
USBIG2024-76-100EA 704 EUR
USBIG2024-76-100 USBIG2024-76-50
Anti-Genistein Mouse Monoclonal Antibody [clone: 5G12]
Antikörper
Recommended dilution: ELISA: ?1:2500

Type: Primary
Antigen: Genistein
Clonality: Monoclonal
Clone: 5G12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR