Anti-Glutamate Transporter EAAT2 Mouse Monoclonal Antibody [clone: 5G109]

Lieferant: US Biological
G4003-91-250 G4003-91-500
USBIG4003-91-250EA 248 EUR
USBIG4003-91-250 USBIG4003-91-500
Anti-Glutamate Transporter EAAT2 Mouse Monoclonal Antibody [clone: 5G109]
Antikörper
Recommended dilution: Immunochemistry: 1:5

Type: Primary
Antigen: Glutamate Transporter EAAT2
Clonality: Monoclonal
Clone: 5G109
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR