Anti-gAcrp30 Goat Polyclonal Antibody (Biotin)

Lieferant: US Biological
G1016-01H-25 G1016-01H-50
USBIG1016-01H-25EA 324 EUR
USBIG1016-01H-25 USBIG1016-01H-50
Anti-gAcrp30 Goat Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml

Type: Primary
Antigen: gAcrp30
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR