Anti-CD24 Rat Monoclonal Antibody [clone: M1/69]

Lieferant: US Biological
C2275-08D-50 C2275-08D-500
USBIC2275-08D-50EA 263 EUR
USBIC2275-08D-50 USBIC2275-08D-500
Anti-CD24 Rat Monoclonal Antibody [clone: M1/69]
Antikörper
Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: M1/69
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR