Anti-CD102 Rat Monoclonal Antibody [clone: 3C4 (MIC2/4)]

Lieferant: US Biological
C2445-39D-500 C2445-39D-50
USBIC2445-39D-500EA 560 EUR
USBIC2445-39D-500 USBIC2445-39D-50
Anti-CD102 Rat Monoclonal Antibody [clone: 3C4 (MIC2/4)]
Antikörper
Type: Primary
Antigen: CD102
Clonality: Monoclonal
Clone: 3C4 (MIC2/4)
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR