Anti-CTBP1 Mouse Monoclonal Antibody [clone: 12L2]

Lieferant: US Biological
C7947-01J-50 C7947-01J-100
USBIC7947-01J-50EA 360 EUR
USBIC7947-01J-50 USBIC7947-01J-100
Anti-CTBP1 Mouse Monoclonal Antibody [clone: 12L2]
Antikörper
Recommended dilution: Western Blot: 1:500

Type: Primary
Antigen: CTBP1
Clonality: Monoclonal
Clone: 12L2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR