Anti-Visual System Homeobox Protein 2 Rabbit Polyclonal Antibody

Lieferant: US Biological
142803 142803-100
USBI142803EA 469 EUR
USBI142803 USBI142803-100
Anti-Visual System Homeobox Protein 2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: Visual System Homeobox Protein 2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR