Anti-4-1 BB Receptor Goat Polyclonal Antibody (Biotin)
Lieferant: US Biological
0004-02A-25 0004-02A-50
USBI0004-02A-25EA 324 EUR
USBI0004-02A-25 USBI0004-02A-50
Anti-4-1 BB Receptor Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: 4-1 BB Receptor
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: IgG
Isotype: Goat
Reactivity: Human,