Anti-ALPI Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-4135S 200-4135-0100
ROCK200-4135SEA 226 EUR
ROCK200-4135S ROCK200-4135-0100
Anti-ALPI Rabbit Polyclonal Antibody
Antikörper
ALKALINE PHOSPHATASE Antibody has been assayed against 1.0 ug of Alkaline Phosphatase [E.coli] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat)

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000

Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR