Anti-Hsf1 Rat Monoclonal Antibody [clone: 10H8]

Lieferant: Cayman Chemical
10011433-100 10011433-25
CAYM10011433-100EA 670 EUR
CAYM10011433-100 CAYM10011433-25
Anti-Hsf1 Rat Monoclonal Antibody [clone: 10H8]
Antikörper
Type: Primary
Antigen: Hsf1
Clonality: Monoclonal
Clone: 10H8
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Bovine, Guinea Pig, Hamster, Human, Mouse, Monkey, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR