Anti-PSD95 Mouse Monoclonal Antibody [clone: 7E3]

Lieferant: Cayman Chemical
10011436-100 10011436-25
CAYM10011436-100EA 683 EUR
CAYM10011436-100 CAYM10011436-25
Anti-PSD95 Mouse Monoclonal Antibody [clone: 7E3]
Antikörper
Type: Primary
Antigen: PSD95
Clonality: Monoclonal
Clone: 7E3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Bovine, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR