Anti-CD19 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 6D5]

Lieferant: BIOLEGEND INC
115537 115538
BLEG115537EA 259 EUR
BLEG115537 BLEG115538
Anti-CD19 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 6D5]
Antikörper
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR