Mouse IgG2b kappa Isotype Control [clone: MPC-11] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)

Lieferant: BIOLEGEND INC
400337 400338
BLEG400337EA 141 EUR
BLEG400337 BLEG400338
Mouse IgG2b kappa Isotype Control [clone: MPC-11] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Antikörper
Type: Control
Antigen: IgG2 β
Clonality: Monoclonal
Clone: MPC-11
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR