Anti-ITGAX Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 3.9]

Lieferant: BIOLEGEND INC
301627 301628
BLEG301627EA 256 EUR
BLEG301627 BLEG301628
Anti-ITGAX Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 3.9]
Antikörper
Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Cynomolgus, Rhesus, African Green Monkey, Baboon, Chimpanzee, Squirrel Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR