Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (Alexa Fluor® 647)

Lieferant: BIOLEGEND INC
400135 400136 400155
BLEG400135EA 123 EUR
BLEG400135 BLEG400136 BLEG400155
Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (Alexa Fluor® 647)
Antikörper
Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR