Anti-CK4, C5, C6, C8, C10, C13, and C18. Mouse Monoclonal Antibody (Unconjugated) [clone: ]

Lieferant: BIOLEGEND INC
628601 628602
BLEG628601EA 149 EUR
BLEG628601 BLEG628602
Anti-CK4, C5, C6, C8, C10, C13, and C18. Mouse Monoclonal Antibody (Unconjugated) [clone: ]
Antikörper
Type: Primary
Antigen: CK4, C5, C6, C8, C10, C13, and C18.
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR