Anti-IgL Rat Monoclonal Antibody (Biotin)

Lieferant: BIOLEGEND INC
407303 407304
BLEG407303EA 123 EUR
BLEG407303 BLEG407304
Anti-IgL Rat Monoclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgL
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope: Light Chain
Host: Rat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR