Anti-IgG2 beta Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: BIOLEGEND INC
406706 406705
BLEG406706EA 304 EUR
BLEG406706 BLEG406705
Anti-IgG2 beta Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG2 beta
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR