Anti-IgE Rat Monoclonal Antibody (PE (Phycoerythrin))

Lieferant: BIOLEGEND INC
406907 406908
BLEG406907EA 174 EUR
BLEG406907 BLEG406908
Anti-IgE Rat Monoclonal Antibody (PE (Phycoerythrin))
Antikörper
Type: Secondary
Antigen: IgE
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR