Anti-IgA Rat Monoclonal Antibody (Biotin)

Lieferant: BIOLEGEND INC
407004 407003
BLEG407004EA 304 EUR
BLEG407004 BLEG407003
Anti-IgA Rat Monoclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgA
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR