Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (APC (Allophycocyanin))

Lieferant: BIOLEGEND INC
400142 400141
BLEG400142EA 245 EUR
BLEG400142 BLEG400141
Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (APC (Allophycocyanin))
Antikörper
Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR