Anti-CD79a Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]

Lieferant: BIOLEGEND INC
333503 333504
BLEG333503EA 193 EUR
BLEG333503 BLEG333504
Anti-CD79a Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]
Antikörper
Type: Primary
Antigen: CD79a
Clonality: Monoclonal
Clone: 0
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR