Anti-IgM Rat Monoclonal Antibody (Brilliant Violet® 421)

Lieferant: BIOLEGEND INC
406517 406518
BLEG406517EA 277 EUR
BLEG406517 BLEG406518
Anti-IgM Rat Monoclonal Antibody (Brilliant Violet® 421)
Antikörper
Type: Primary
Antigen: IgM
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR